Order Entry
Spain
ContactUsLinkComponent
trans-1,2-Dimethylcyclohexane ≥99.0% (by GC)
trans-1,2-Dimethylcyclohexane ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
trans-1,2-Dimethylcyclohexane ≥99.0% (by GC)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

Frequently Bought Together