4-Chlorobenzoic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: p-Chlorobenzoic acid, Chlorodracyclic acid, Chlorodracylic acid
CAS Number:74-11-3
:
Formula:C₇H₅ClO₂
MDL Number:MFCD00002531
Boiling Pt:287
Molecular Weight:156.57 g/mol
Melting Pt:123
Flash Pt:238
Merck Index:14,02125
EINECS:200-805-9
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:235.0 to 245.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together