Phenyl benzoate ≥99.0% (by GC)
: TCI
: Benzoic acid phenyl ester
:93-99-2
:
:C₁₃H₁₀O₂
:MFCD00003072
:299
:198.22 g/mol
:14,07275
:202-293-2
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:68.0 to 72.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together