Order Entry
Spain
ContactUsLinkComponent
 
(±)-2,3-Dimethyl-2-hexanol ≥99.0% (by GC)
  TCIAD1967-5ML
 :  TCI
 :  
undefined
(±)-2,3-Dimethyl-2-hexanol ≥99.0% (by GC)
  TCIAD1967-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 : 2,3-Dimethyl-2-hexanol
 :
 :130.23 g/mol
 :1987
 :GHS02
 :19550-03-9
 :C₈H₁₈O
 :MFCD00021806
 :69°C
 :-45
 :3 III