2-Methyl-2-butanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: tert-Amyl alcohol, tert-Pentyl alcohol, tert-Pentanol
CAS Number:75-85-4
:
Formula:C₅H₁₂O
MDL Number:MFCD00004478
Boiling Pt:102°C
Molecular Weight:88.15 g/mol
Melting Pt:–8°C
Flash Pt:41°C
ADR:3 III
Merck Index:14,07140
EINECS:200-908-9
UN:1105
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together