Order Entry
Spain
ContactUsLinkComponent
Cesium fluoride
Cesium fluoride
Supplier:  TCI
Certificates
undefined
Cesium fluoride
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White powder to crystal
  • Purity:
    min. 99.0 %

Specification Test Results

Frequently Bought Together