Askina® Elast fine Fixation Bandage
Supplier: B. Braun
Certificates
About this item
Elastic fixation bandage made from 54% polyamide and 46% viscose for securing wound dressings and splints and for light compression bandages.
- Approx. 140% lengthways elasticity
- Secure, long-lasting hold thanks to supple and elastic material
- High stability thanks to woven edge
DIN 61634
Frequently Bought Together