(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-(4-Hydroxy-3,5-dimethoxyphenyl)acetic acid, 4-Hydroxy-3,5-dimethoxyphenylacetic acid
CAS Number:4385-56-2
:
Formula:C₁₀H₁₂O₅
MDL Number:MFCD00016834
Molecular Weight:212.2 g/mol
Melting Pt:127…131 °C
EINECS:224-494-4
:GHS06
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together