Anti-PSMB4 Goat Polyclonal Antibody
Catalog # ANTIA84866-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:26 kDa prosomal protein
- Antigen symbol:PSMB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:HSN3|HsBPROS26|Macropain beta chain|Multicatalytic endopeptidase complex beta chain|Proteasome beta chain|PROS-26|Proteasome (prosome macropain) subunit beta type 4|HN3|Proteasome beta 4 subunit
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-NRFQIATVTE
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PSMB4 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to PSMB4 for ELISA and WB with samples derived from human, mouse and rat.
- Validated applications: ELISA, W
Type: Primary
Antigen: PSMB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat