Order Entry
Spain
ContactUsLinkComponent
Anti-CD59 Rabbit Recombinant Antibody [clone: MACIF/2867R]
Anti-CD59 Rabbit Recombinant Antibody [clone: MACIF/2867R]
Catalog # ANTIA250742-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD59 Rabbit Recombinant Antibody [clone: MACIF/2867R]
Catalog # ANTIA250742-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    1F5 antigen
  • Antigen symbol:
    CD59
  • Clonality:
    Recombinant
  • Clone:
    MACIF/2867R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13987
  • Antigen synonyms:
    CD59 antigen|CD59 antigen complement regulatory protein|1F5|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|16.3A5|20 kDa homologous restriction factor|CD59 antigen p18 20
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    20 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CD59 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [MACIF/2867R] antibody to CD59 for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD59
Clonality: Recombinant
Clone: MACIF/2867R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human