Anti-TFB2M Rabbit Polyclonal Antibody
PRSI55-058
New Product
- Antibody type:Primary
- Antigen name:Dimethyladenosine transferase 2, mitochondrial
- Antigen symbol:TFB2M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Amino Acid Range = 368 - 396
- Form:Liquid
- Gene ID:Q9H5Q4
- Antigen synonyms:TFB2M
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:45 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This TFB2M antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 368 - 396 amino acids from the C-terminal region of human TFB2M
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For IHC-P starting dilution is: 1:50~100 For WB starting dilution is: 1:1000.
Type: Primary
Antigen: TFB2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 368-396
Host: Rabbit
Isotype:
Reactivity: Human