Order Entry
Spain
ContactUsLinkComponent
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 97%
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 97%
Catalog # APOSOR912500-5G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2-Amino-3-(4-cyanophenyl)propanoic acid 97%
Catalog # APOSOR912500-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Cyano-D-phenylalanine, H-4-Cyano-D-Phe-OH, H-D-Phe(4-CN)-OH
 :
 :190.202 g/mol
Storage Temperature:Ambient
CAS Number:263396-44-7
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD00270364