Order Entry
Spain
ContactUsLinkComponent
Methyl adipoyl chloride 96%
Methyl adipoyl chloride 96%
Catalog # APOSOR4232-25G
Supplier:  Apollo Scientific
CAS Number:  
Methyl adipoyl chloride 96%
Catalog # APOSOR4232-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Methyl 6-chloro-6-oxohexanoate
 :
 :178.62 g/mol
Boiling Pt:76 °C/0.8mmHg
UN:3265
 :GHS05
Storage Temperature:Ambient
CAS Number:35444-44-1
Formula:C₇H₁₁ClO₃
MDL Number:MFCD00013661
Density:1.05 g/cm³ (25 °C)
ADR:8 II