Order Entry
Spain
ContactUsLinkComponent
 
2,3-Dimethyloctane
Catalog # APOSOR2371-1G
Supplier:  Apollo Scientific
CAS Number:  
2,3-Dimethyloctane
Catalog # APOSOR2371-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-Isopropylheptane
 :
 :142.282 g/mol
UN:3295
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:7146-60-3
Formula:C₁₀H₂₂
MDL Number:MFCD00048875
ADR:3 III