Order Entry
Spain
Orders LinkContactUsLinkComponent
1-Hexyl-4-iodobenzene ≥98%

1-Hexyl-4-iodobenzene ≥98%

1-Hexyl-4-iodobenzene ≥98%
Certificates
1-Hexyl-4-iodobenzene ≥98%
1-Hexyl-4-iodobenzene ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 62150-34-9 Molecular Formula: C12H17I Formula Weight: 288.16 Flash Point: 120°(248°F) Refractive Index: 1.5530 Boiling Point: 122-125°/3mm MDL No.: MFCD07777097

  • CAS Number:62150-34-9
  • Formula:C₁₂H₁₇I
  • MDL Number:MFCD07777097
  • Molecular Weight:288.17 g/mol
  • Boiling Pt:122…125 °C (3 mmHg)
  • Flash Pt:>120 °C (248 °F)
  • Storage Temperature:Ambient
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.