Mefenpyr-diethyl ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
Synonyms: Diethyl 1-(3,4-Dichlorophenyl)-5-methyl-2-pyrazoline-3,5-dicarboxylate, 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic acid Diethyl Ester, Diethyl 1-(2,4-dichlorophenyl)-5-methyl-4,5-dihydro-1H-pyrazole-3,5-dicarboxylate, Diethyl 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate
CAS Number:135590-91-9
:
Formula:C₁₆H₁₈Cl₂N₂O₄
MDL Number:MFCD09753375
Molecular Weight:373.23 g/mol
UN:3077
ADR:9 III
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:50.0 to 54.0 deg-C
Specification Test Results
Frequently Bought Together