Order Entry
Spain
ContactUsLinkComponent
 
Adenine
Catalog # EDQMA0230000
Supplier:  EDQM
CAS Number:  
Adenine
Catalog # EDQMA0230000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine
 :
 :135.13 g/mol
EINECS:200-796-1
UN:2811
 :GHS06
Storage Temperature:Ambient
CAS Number:73-24-5
Formula:C₅H₅N₅
MDL Number:MFCD00041790
Density:0.99741 g/cm³ (25 °C)
ADR:6.1 III
Merck Index:13,00152