2-Fluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Fluorobenzene-1-sulphonyl chloride
CAS Number:2905-21-7
:
Formula:C₆H₄ClFO₂S
MDL Number:MFCD00042285
Boiling Pt:246…247 °C
Melting Pt:27…32 °C
Density:1.470
Molecular Weight:194.61 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together