Benzocaine (ethyl-4-aminobenzoate) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ethyl 4-aminobenzoate, Aethoform
CAS Number:94-09-7
:
Formula:C₉H₁₁NO₂
MDL Number:MFCD00007892
Boiling Pt:310 °C
Molecular Weight:165.19 g/mol
Melting Pt:88…92 °C
Flash Pt:>110 °C (230 °F)
Merck Index:13,01087
EINECS:202-303-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberALFAA12754.30_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together