Potassium dichromate 1/60 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 191005X
Supplier: VWR Chemicals
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.01663 - 0.01670 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:294.18 g/mol
EINECS:231-906-6
UN:3287
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367
Boiling Pt:500
Melting Pt:398
ADR:6.1 III
Merck Index:12,07790