Guanidinium hydrochloride 8 mol/l in aqueous solution
Catalog # PIER24115
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:200 mL
- Seq:2530
- Environmentally Preferable:
Specifications
About this item
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
:
:95.53 g/mol
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
CAS Number:50-01-1
Formula:NH₂C(=NH)NH₂·HCl
MDL Number:MFCD00013026
Melting Pt:185
Merck Index:13,04578