Restriction enzymes, NdeI, Fermentas
Supplier: Thermo Fisher Scientific
Certificates
About this item
NdeI restriction enzyme recognises CA^TATG sites and cuts at 37 °C in O buffer (Isoschizomers: FauNDI).
- Used for molecular cloning, restriction site mapping, genotyping, southern blotting, RFLP and SNP.
Superior quality, stringent quality control and industry leading manufacturing process
Convenient colour-coded five buffer system
BSA premixed in reaction buffers
Wide selection of restriction endonuclease specificities
Delivery: Supplied with universal Tango buffer for double-digestions.
Specifications
- Application:
- Source:
- Environmentally Preferable:
Specifications
Frequently Bought Together