Order Entry
Spain
ContactUsLinkComponent
 
Anti-EDG-7 Rabbit Polyclonal Antibody [clone: P324]
  BWRLBS2791
Anti-EDG-7 Rabbit Polyclonal Antibody [clone: P324]
  BWRLBS2791

 

  • Antibody type:
    Primary
  • Antigen symbol:
    EDG-7
  • Clonality:
    Polyclonal
  • Clone:
    P324
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 289-342 of Human EDG-7.

EDG-7 (P324) pAb detects endogenous levels of EDG-7 protein.

Type: Primary
Antigen: EDG-7
Clonality: Polyclonal
Clone: P324
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human