Order Entry
Spain
ContactUsLinkComponent
HEV-ORF3
HEV-ORF3
Catalog # BIRBORB97973-100
Supplier:  Biorbyt
CAS Number:  
HEV-ORF3
Catalog # BIRBORB97973-100
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HEV-ORF3
  • Purity:
    Greater than 95% by SDS-PAGE
  • Formulation:
    Liquid. Supplied in 20 mM Tris-HCl pH 8.0, 10 mMmercaptoethanol and 8M urea.
  • Tested applications:
    ELISA , WB

Specifications

About this item