Anti-ERV3-1 Rabbit Polyclonal Antibody
Catalog # BSBTRP1091
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:endogenous retrovirus group 3, member 1
- Antigen symbol:ERV3-1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2086
- Antigen synonyms:ERV3|HERV-R|HERVR|ERV-R|ERVR|envR
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Endogenous retrovirus group 3 member 1 Env polyprotein(ERV3-1) detection. Tested with WB in Human.
Type: Primary
Antigen: ERV3-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human