Order Entry
Spain
ContactUsLinkComponent
Anti-MAPK11 Rabbit Polyclonal Antibody
Anti-MAPK11 Rabbit Polyclonal Antibody
  ABNOPAB0372
 :  Abnova
Anti-MAPK11 Rabbit Polyclonal Antibody
  ABNOPAB0372
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 11
  • Antigen symbol:
    MAPK11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5600
  • Antigen synonyms:
    PRKM11|SAPK2|p38-2|P38B|P38BETA2|SAPK2B|p38Beta
  • Amino acid number:
    39 to 59
  • Storage buffer:
    PBS (0,2% gelatin, 0,05% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 39-59 of human MAPK11.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of MAPK11.

Type: Primary
Antigen: MAPK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat