Anti-BNIP1 Goat Polyclonal Antibody
ABNOPAB7347
: Abnova
- Antibody type:Primary
- Antigen name:BCL2/adenovirus E1B 19kDa interacting Protein 1
- Antigen symbol:BNIP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:662
- Antigen synonyms:NIP1|TRG-8|SEC20
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DLLRQRKTTKESLAQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human BNIP1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Goat polyclonal antibody raised against synthetic peptide of BNIP1.
Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.5-1.5 ?g/ml
Type: Primary
Antigen: BNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human