Anti-TLR6 Rabbit Polyclonal Antibody
ABNOPAB9734
: Abnova
- Antibody type:Primary
- Antigen name:Toll-like Receptor 6
- Antigen symbol:TLR6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:10333
- Antigen synonyms:CD antigen 286|CD286|Toll-like receptor 6|TLR6
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human TLR6.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of TLR6.
Type: Primary
Antigen: TLR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse