Anti-ABCB5 Rabbit Polyclonal Antibody
100 μl
Catalog # ORIGTA323978
Supplier: OriGene
Jump to:
Product Details & Documents
Recommended Dilutions: ELISA: 1:500-2000; Immunochemistry: 1:10-50
Type: Primary
Antigen: ABCB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
TA323978
Datasheet
Specifications
- Size:100 μl
- Clonality:Polyclonal
- Purification:purified by antigen affinity chromatography
- Antibody type:Primary
- Form:Liquid
- Amino acid number:corresponding to a region between amino acids 563 to 812
- Isotype:IgG
- Storage temperature:–20 °C
- Host:Rabbit
- Cross adsorption:No
- Shipping temperature:–20 °C
- Antigen name:ATP Binding Cassette, sub-family B, member 5
- Cat. No.:ORIGTA323978
- Reactivity:Human|Rat|Mouse
- Immunogen:Fusion protein corresponding to a region derived from 563-812 amino acids of Human ATP-binding cassette sub-family B member 5
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:ABCB5
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



