Order Entry
Spain
ContactUsLinkComponent
Multi-parameter meters, bench, HQd (HQ430d / HQ440d)
Multi-parameter meters, bench, HQd (HQ430d / HQ440d)
Supplier:  Hach
Certificates
undefined
Multi-parameter meters, bench, HQd (HQ430d / HQ440d)
Supplier:  Hach

Specifications

  • Catalog No:
  • HACHHQ430D9800002
  • 665-0381
  • HACHHQ440D9800002
  • 665-0382
  • Model:
  • HQ430d
  • HQ430d
  • HQ440d
  • HQ440d
  • pH range:
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • pH resolution:
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • pH calibration:
  • Colour-coded: 4,01/7,00/10,01; IUPAC: 1,679/4,005/7,000/10,012/12,45; DIN: 1,09/4,65/9,23; user-defined custom buffer sets
  • Colour-coded: 4,01/7,00/10,01; IUPAC: 1,679/4,005/7,000/10,012/12,45; DIN: 1,09/4,65/9,23; user-defined custom buffer sets
  • Colour-coded: 4,01/7,00/10,01; IUPAC: 1,679/4,005/7,000/10,012/12,45; DIN: 1,09/4,65/9,23; user-defined custom buffer sets
  • Colour-coded: 4,01/7,00/10,01; IUPAC: 1,679/4,005/7,000/10,012/12,45; DIN: 1,09/4,65/9,23; user-defined custom buffer sets
  • mV range:
  • ±1500
  • ±1500
  • ±1500
  • ±1500
  • mV resolution:
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • 0,1/ 0,01/ 0,001
  • mV accuracy:
  • ±0,1
  • ±0,1
  • ±0,1
  • ±0,1
  • ISE range:
  • Electrode specific
  • Electrode specific
  • Electrode specific
  • Electrode specific
  • ISE resolution:
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • 0,1/0,01/0,001
  • ISE accuracy:
  • ±0,1 mV
  • ±0,1 mV
  • ±0,1 mV
  • ±0,1 mV
  • Conductivity range:
  • 0,01 μS/cm - 200 mS/cm
  • 0,01 μS/cm - 200 mS/cm
  • 0,01 μS/cm - 200 mS/cm
  • 0,01 μS/cm - 200 mS/cm
  • Conductivity resolution:
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • 5 digits with 2 digits after decimal point
  • Conductivity accuracy:
  • ±0,5% (1 μS/cm - 200 mS/cm)
  • ±0,5% (1 μS/cm - 200 mS/cm)
  • ±0,5% (1 μS/cm - 200 mS/cm)
  • ±0,5% (1 μS/cm - 200 mS/cm)
  • Conductivity calibration:
  • Demal (1D / 0,1D / 0,01D); Molar (0,1M / 0,01M / 0,001M); NaCl (0,05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
  • Demal (1D / 0,1D / 0,01D); Molar (0,1M / 0,01M / 0,001M); NaCl (0,05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
  • Demal (1D / 0,1D / 0,01D); Molar (0,1M / 0,01M / 0,001M); NaCl (0,05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
  • Demal (1D / 0,1D / 0,01D); Molar (0,1M / 0,01M / 0,001M); NaCl (0,05%/25 µS/cm/1000 µS/cm/18 mS/cm); standard sea water; user defined
  • Resistivity range:
  • 2,5 Ωcm - 49 MΩcm
  • 2,5 Ωcm - 49 MΩcm
  • 2,5 Ωcm - 49 MΩcm
  • 2,5 Ωcm - 49 MΩcm
  • Resistivity accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • Salinity range:
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • 0 - 42 g/kg
  • Salinity resolution:
  • 0,01 ppt
  • 0,01 ppt
  • 0,01 ppt
  • 0,01 ppt
  • Salinity accuracy:
  • ±0,1 mg/L (for range <8 mg/L)
  • ±0,1 mg/L (for range <8 mg/L)
  • ±0,1 mg/L (for range <8 mg/L)
  • ±0,1 mg/L (for range <8 mg/L)
  • TDS range:
  • 0,0 - 50,0 g/L
  • 0,0 - 50,0 g/L
  • 0,0 - 50,0 g/L
  • 0,0 - 50,0 g/L
  • TDS resolution:
  • max. 3 digits
  • max. 3 digits
  • max. 3 digits
  • max. 3 digits
  • TDS accuracy:
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • ±0,5%
  • DO concentration:
  • 0,00 - 20,0 mg/L
  • 0,00 - 20,0 mg/L
  • 0,00 - 20,0 mg/L
  • 0,00 - 20,0 mg/L
  • DO concentration resolution:
  • 0,01 mg/L
  • 0,01 mg/L
  • 0,01 mg/L
  • 0,01 mg/L
  • Temperature range:
  • −10...+110 °C
  • −10...+110 °C
  • −10...+110 °C
  • −10...+110 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ± 0,3 °C
  • ± 0,3 °C
  • ± 0,3 °C
  • ± 0,3 °C
  • Temperature compensation:
  • Automatic, off, manual
  • Automatic, off, manual
  • Automatic, off, manual
  • Automatic, off, manual
  • Display:
  • Backlit graphical LCD
  • Backlit graphical LCD
  • Backlit graphical LCD (with simultaneous readings from two electrodes)
  • Backlit graphical LCD (with simultaneous readings from two electrodes)
  • Power supply:
  • 230 V with power supply or 4 × AA Alkaline batteries
  • 230 V with power supply or 4 × AA Alkaline batteries
  • 230 V with power supply or 4 × AA Alkaline batteries
  • 230 V with power supply or 4 × AA Alkaline batteries
  • Data logging capacity:
  • 500 results
  • 500 results
  • 500 results
  • 500 results
  • Connections:
  • 1 × M12 digital for IntelliCAL™ electrodes
  • 1 × M12 digital for IntelliCAL™ electrodes
  • 2 × M12 digital for IntelliCAL™ electrodes
  • 2 × M12 digital for IntelliCAL™ electrodes
  • Outputs:
  • USB
  • USB
  • USB
  • USB
  • W×D×H:
  • 175×234×86
  • 175×234×86 mm
  • 175×234×86
  • 175×234×86 mm
  • Weight:
  • 0,85 kg
  • 0,85 kg
  • 0,85 kg
  • 0,85 kg

Specifications

Related Information

pH electrodes

Product Information

Frequently Bought Together