Anti-PSMD13 Rabbit Polyclonal Antibody (Biotin)
USBIP9078-64A-BIOT
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) 26S subunit, non-ATPase 13
- Antigen symbol:PSMD13
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: PSMD13 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 13)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human