Anti-Furin Convertase, Region 2 Mouse Monoclonal Antibody [clone: 6D404]
Catalog # USBIF9201-11
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Furin Convertase, Region 2
- Clonality:Monoclonal
- Clone:6D404
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Format:ascites
- Form:liquid
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunochemistry: 1:200-1:400; Immunoprecipitation: 1:200; Western Blot: 1:1000
Type: Primary
Antigen: Furin Convertase, Region 2
Clonality: monoclonal
Clone: 6D404
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse