Anti-CHAT Rabbit Polyclonal Antibody
Catálogo # USBIC5057-10M
Proveedor: US Biological
Especificaciones
- Antibody type:Primary
- Antigen name:Choline Acetyltransferase
- Antigen symbol:ChAT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish
- Environmentally Preferable:
- Epitope:C-terminal
- Form:liquid
- Storage buffer:40mMMOPS, pH 7,5, 0,1% BSA, 0,05% sodium azide, 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:250 µl
- Pk:250 µl
Especificaciones
Acerca de este artículo
Recommended dilution: ELISA: 1:5000-1:20000
Type: Primary
Antigen: CHAT (choline O-acetyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish