Anti-GEMIN7 Mouse Monoclonal Antibody [clone: 2E2]
USBI127270
- Antibody type:Primary
- Antigen name:Gem (nuclear organelle) associated protein 7
- Antigen symbol:GEMIN7
- Clonality:Monoclonal
- Clone:2E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Sequence:MQTPVNIPVPVLRLPRGPDGFSRGFAPDGRRAPLRPEVPEIQECPIAQESLESQEQRARAALRERYLRSLLAMVGHQVSFTLHEGVRVAAHFGATDLDVANFYVSQLQTPIGVQAEALLRCSDIISYTFKP
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: GEMIN7 (gem (nuclear organelle) associated protein 7)
Clonality: monoclonal
Clone: 200
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human