Anti-DYM Mouse Monoclonal Antibody [clone: 1C2]
USBI126087
- Antibody type:Primary
- Antigen name:Dymeclin
- Antigen symbol:DYM
- Clonality:Monoclonal
- Clone:1C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Amino acid number:343 to 431
- Storage buffer:PBS, pH 7,2.
- Sequence:QTSDQATLLLYTLLHQNSNIRTYMLARTDMENLVLPILEILYHVEERNSHHVYMALIILLILTEDDGFNRSIHEVILKNITWYSERVL
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: DYM (dymeclin)
Clonality: monoclonal
Clone: 1C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human