Anti-ATG9A Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ATG9A (autophagy related 9A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNB110-56893SS
- NOVUNB110-56893
- Antigen symbol:
- ATG9A
- ATG9A
- Antigen name:
- Autophagy Related 9A
- Autophagy Related 9A
- Antigen synonyms:
- APG9L1|ATG9A|MGD3208|APG9-like 1|mATG9
- APG9L1|ATG9A|MGD3208|APG9-like 1|mATG9
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 79065
- 79065
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together