Order Entry
Spain
ContactUsLinkComponent
 
Anti-HIF-1 beta Rabbit Polyclonal Antibody
 
undefined
Anti-HIF-1 beta Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB100-110
  • NOVUNB100-110SS
  • Antigen symbol:
  • HIF1B
  • HIF1B
  • Antigen name:
  • HIF1 beta
  • HIF1 beta
  • Antigen synonyms:
  • class E basic helix-loop-helix protein 2|HIF-1-beta|HIF1BETA|HIF-1beta|dioxin receptor|hypoxia-inducible factor 1|nuclear translocator|bHLHe2|TANGO|HIF1B|beta subunit|HIF1-beta
  • class E basic helix-loop-helix protein 2|HIF-1-beta|HIF1BETA|HIF-1beta|dioxin receptor|hypoxia-inducible factor 1|nuclear translocator|bHLHe2|TANGO|HIF1B|beta subunit|HIF1-beta
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 405
  • 405
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • ChIP:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together