Anti-Lom-TK 2 Rabbit Polyclonal Antibody
BIRBORB122517-100
: Biorbyt
- Antibody type:Primary
- Antigen symbol:LOM-TK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide.
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Lom-TK 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: