Anti-AZIN1 Rabbit Polyclonal Antibody
ABNOH00051582-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Antizyme Inhibitor 1
- Antigen symbol:AZIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:51582
- Antigen synonyms:AZIA1|OAZIN|ODC1L|OAZI|AZI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AZIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human