Anti-NMT2 Mouse Monoclonal Antibody [clone: 2E12-4B5]
ABNOH00009397-M01
: Abnova
- Antibody type:Primary
- Antigen name:N-myristoyltransferase 2
- Antigen symbol:NMT2
- Clonality:Monoclonal
- Clone:2E12-4B5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9397
- Antigen synonyms:Peptide N myristoyltransferase 2|Glycylpeptide N tetradecanoyltransferase 2|Type II N myristoyltransferase|NMT 2|NMT2_HUMAN.|Myristoyl CoA:protein N myristoyltransferase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: NMT2
Clonality: Monoclonal
Clone: 2E12-4B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human