Anti-UAP1 Mouse Polyclonal Antibody
ABNOH00006675-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Udp-N-Acetylglucosamine Pyrophosphorylase 1
- Antigen symbol:UAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6675
- Antigen synonyms:AGX2|SPAG2|AGX|AGX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: UAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human