Anti-ADI1 Rabbit Polyclonal Antibody
AVIVARP66315_P050
- Antibody type:Primary
- Antigen name:Acireductone dioxygenase 1
- Antigen symbol:ADI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:55256
- Antigen synonyms:MTCBP1|Ni-ARD|ARD|APL1|HMFT1638|Fe-ARD|mtnD|SIPL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ADI1 (acireductone dioxygenase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human