Anti-CTSV Rabbit Polyclonal Antibody
AVIVARP61284_P050
- Antibody type:Primary
- Antigen name:cathepsin V
- Antigen symbol:CTSV
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1515
- Antigen synonyms:CATL2|CTSU|CTSL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CTSV (cathepsin V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human