Anti-CHI3L1 Rabbit Polyclonal Antibody
Catalog # AVIVARP51929_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Chitinase 3-like 1
- Antigen symbol:CHI3L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:1116
- Antigen synonyms:ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CHI3L1 (chitinase 3-like 1 (cartilage glycoprotein-39))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: