Hydrophobic interaction chromatography resins, Toyopearl®, TSK-GEL®
These are bulk media products which contain three or four different ligand types offered for a chromatography mode.
Physical properties of resins in different buffer systems can be identified by slurry and slurry mechanics, resin handling during column packing, mechanical strength relative to other resin backbones and degree of compressibility.
It is used to determine the resin's dynamic binding capacity, selectivity, column efficiency, operating conditions by developmental scientist or engineer.
- Environmentally Preferable:
Frequently Bought Together