Order Entry
Denmark
ContactUsLinkComponent
 
Cyclododecene cis- and trans-mixture ≥95.0% (by GC)
  TCIAC0464-25G
 :  TCI
 :  
undefined
Cyclododecene cis- and trans-mixture ≥95.0% (by GC)
  TCIAC0464-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

 

 

 :
 :166.31 g/mol
 :216-117-7
 :1501-82-2
 :C₁₂H₂₂
 :MFCD00003721
 :232
 :-9
 :94°C