Diethyl cyclobutane-1,1-dicarboxylate ≥97.0% (by GC)
: TCI
:3779-29-1
:
:C₁₀H₁₆O₄
:MFCD00019261
:226°C
:200.24 g/mol
:89°C
:223-239-4
- Environmentally Preferable:
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Frequently Bought Together