Order Entry
Denmark
ContactUsLinkComponent
Chlorotriphenylgermane ≥97.0% (by GC)
Chlorotriphenylgermane ≥97.0% (by GC)
Catalog # TCIAT1162-1G
Supplier:  TCI
CAS Number:  
undefined
Chlorotriphenylgermane ≥97.0% (by GC)
Catalog # TCIAT1162-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Yellow powder to crystal
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

Synonyms: Triphenylgermanium chloride, Chlorotriphenylgermane
 :
 :339.38 g/mol
EINECS:216-617-5
 :GHS07
CAS Number:1626-24-0
Formula:C₁₈H₁₅ClGe
MDL Number:MFCD00000461