2,6-Dimethoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:23112-96-1
:
Formula:C₈H₁₁BO₄
MDL Number:MFCD01318987
Molecular Weight:181.98 g/mol
Melting Pt:113°C
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together