Pycnometer
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in stopper
- Good chemical resistance
- Good temperature resistance
: DIN ISO 3507
Frequently Bought Together
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.